Assay Design



Gene Transcripts

Gene Symbol
FGF8
Entrez Gene ID
396313
Gene Name
fibroblast growth factor 8
Gene Aliases
FGF-8
Chromosome Location
Chr.6: 23669271 - 23677126 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012767.1 NP_001012785.1 4 - 5 363 129
XM_015288595.2 XP_015144081.1 4 - 5 545 129
GenBank mRNA FJ882410.1 - 4 - 5 363 129
U55189.1 - 4 - 5 363 129

View Full Product Details