Assay Design



Gene Transcripts

Gene Symbol
YTHDF1
Entrez Gene ID
419238
Gene Name
YTH N6-methyladenosine RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 8701741 - 8714334 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012833.1 NP_001012851.1 1 - 2 384 96
XM_015296384.2 XP_015151870.2 1 - 2 35 96
GenBank mRNA AJ719437.1 - 1 - 2 376 96
AJ851651.1 - 1 - 2 384 96
BX933801.1 - 1 - 2 122 96

View Full Product Details