Assay Design



Gene Transcripts

Gene Symbol
EEF2KMT
Entrez Gene ID
416392
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
FAM86A
Chromosome Location
Chr.14: 13793040 - 13797471 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030627.1 NP_001025798.1 2 - 3 304 89
GenBank mRNA AJ720764.1 - 2 - 3 304 89

View Full Product Details