Assay Design



Gene Transcripts

Gene Symbol
NSFL1C
Entrez Gene ID
419268
Gene Name
NSFL1 cofactor
Gene Aliases
-
Chromosome Location
Chr.20: 9807241 - 9814899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030859.1 NP_001026030.1 5 - 6 536 64
XM_015296393.2 XP_015151879.1 5 - 6 766 64
GenBank mRNA AJ720274.1 - 5 - 6 536 64

View Full Product Details