Assay Design



Gene Transcripts

Gene Symbol
LIN7C
Entrez Gene ID
421608
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Gene Aliases
LIN7B
Chromosome Location
Chr.5: 3937290 - 3949893 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031067.1 NP_001026238.1 2 - 3 184 91
XM_015286169.2 XP_015141655.2 2 - 3 192 91
XM_015286170.2 XP_015141656.1 2 - 3 267 91
GenBank mRNA AJ851475.1 - 2 - 3 184 91

View Full Product Details