Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031222.1 | NP_001026393.1 | 3 - 4 | 456 | 76 |
XM_015288131.2 | XP_015143617.2 | 4 - 5 | 76 | ||
XM_015288132.2 | XP_015143618.2 | 3 - 4 | 76 | ||
XM_015288133.2 | XP_015143619.2 | 4 - 5 | 76 | ||
XM_015288134.2 | XP_015143620.2 | 4 - 5 | 76 | ||
XM_015288135.2 | XP_015143621.2 | 3 - 4 | 76 | ||
GenBank mRNA | AJ720344.1 | - | 3 - 4 | 456 | 76 |