Assay Design



Gene Transcripts

Gene Symbol
KRT13
Entrez Gene ID
408040
Gene Name
keratin 13
Gene Aliases
KRT15
Chromosome Location
Chr.27: 7361940 - 7366365 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001312.2 NP_001001312.2 3 - 4 817 66
GenBank mRNA AY574986.1 - 4 - 5 719 66
CN227144.1 - 1 - 2 79 66

View Full Product Details