Assay Design



Gene Transcripts

Gene Symbol
FGF13
Entrez Gene ID
414831
Gene Name
fibroblast growth factor 13
Gene Aliases
FHF-2
Chromosome Location
Chr.4: 4747075 - 4962390 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001743.1 NP_001001743.1 3 - 4 447 121
XM_015278206.2 XP_015133692.1 4 - 5 753 121
XM_015278207.2 XP_015133693.1 3 - 4 853 121
XM_015278209.2 XP_015133695.1 3 - 4 368 121
GenBank mRNA AF108755.1 - 3 - 4 605 121
AF199610.1 - 3 - 4 447 121
AF199611.1 - 3 - 4 721 121

View Full Product Details