Assay Design
Gene Transcripts
- Gene Symbol
- MRPS30
- Entrez Gene ID
-
395781
- Gene Name
- mitochondrial ribosomal protein S30
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Z: 14183679 - 14189220 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204939.1 |
NP_990270.1 |
3 - 4 |
986 |
89 |
GenBank mRNA |
AF029071.1 |
- |
3 - 4 |
986 |
89 |
View Full Product Details