Assay Design



Gene Transcripts

Gene Symbol
KRT24
Entrez Gene ID
395861
Gene Name
keratin 24
Gene Aliases
GK-19, KRT19, keratin-19
Chromosome Location
Chr.27: 7369277 - 7373939 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205009.2 NP_990340.2 5 - 6 1104 99
GenBank mRNA AB016281.1 - 5 - 6 1104 99
BX259386.3 - 2 - 3 300 99
CR390877.1 - 2 - 3 135 99

View Full Product Details