Assay Design
Gene Transcripts
- Gene Symbol
- SPI1
- Entrez Gene ID
-
395879
- Gene Name
- Spi-1 proto-oncogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 23185094 - 23206769 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205023.1 |
NP_990354.1 |
3 - 4 |
556 |
84 |
GenBank mRNA |
Y12225.1 |
- |
- |
556 |
84 |
View Full Product Details