Assay Design



Gene Transcripts

Gene Symbol
BPIFB2
Entrez Gene ID
395882
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIFB7, G2, TENP
Chromosome Location
Chr.20: 10296177 - 10301937 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205026.1 NP_990357.1 6 - 7 527 79
XM_015296335.2 XP_015151821.1 6 - 7 547 79
XM_025142176.1 XP_024997944.1 6 - 7 554 79
GenBank mRNA AB219157.1 - 5 - 6 481 79
AB219158.1 - 5 - 6 481 79
AB219159.1 - 5 - 6 481 79
AB219160.1 - 5 - 6 481 79
AF029841.1 - 6 - 7 527 79
HG007958.1 - 6 - 7 560 79
LC144559.1 - 5 - 6 481 79
LC144560.1 - 5 - 6 481 79
LC144561.1 - 5 - 6 481 79
LC144562.1 - 5 - 6 481 79
LC144563.1 - 5 - 6 481 79
LC144564.1 - 5 - 6 481 79
LC144565.1 - 5 - 6 481 79
LC144566.1 - 5 - 6 481 79
LC144567.1 - 5 - 6 481 79
LC144568.1 - 5 - 6 481 79
LC144569.1 - 5 - 6 481 79
LC144570.1 - 5 - 6 481 79
LC144571.1 - 5 - 6 481 79
LC144572.1 - 5 - 6 481 79
LC144573.1 - 5 - 6 481 79
LC144574.1 - 5 - 6 481 79
LC144575.1 - 5 - 6 481 79
LC144576.1 - 5 - 6 481 79
LC144577.1 - 5 - 6 481 79
LC144578.1 - 5 - 6 481 79
LC144579.1 - 5 - 6 481 79
LC144580.1 - 5 - 6 481 79
LC144581.1 - 5 - 6 481 79
LC144582.1 - 5 - 6 481 79
LC144583.1 - 5 - 6 481 79
LC144584.1 - 5 - 6 481 79
LC144585.1 - 5 - 6 481 79
LC144586.1 - 5 - 6 481 79
LC144587.1 - 5 - 6 481 79
LC144588.1 - 5 - 6 481 79
LC144589.1 - 5 - 6 481 79
LC144590.1 - 5 - 6 481 79
LC144591.1 - 5 - 6 481 79
LC144592.1 - 5 - 6 481 79
LC144593.1 - 5 - 6 481 79
LC144594.1 - 5 - 6 481 79
LC144595.1 - 5 - 6 481 79
LC144596.1 - 5 - 6 481 79
LC144597.1 - 5 - 6 481 79
LC144598.1 - 5 - 6 481 79
LC144599.1 - 5 - 6 481 79
LC144600.1 - 5 - 6 481 79
LC144601.1 - 5 - 6 481 79
LC144602.1 - 5 - 6 481 79
LC144603.1 - 5 - 6 481 79
LC144604.1 - 5 - 6 481 79
LC144605.1 - 5 - 6 481 79
LC144606.1 - 5 - 6 481 79
LC144607.1 - 5 - 6 481 79
LC144608.1 - 5 - 6 481 79

View Full Product Details