Assay Design
Gene Transcripts
- Gene Symbol
- GABRE
- Entrez Gene ID
-
396173
- Gene Name
- gamma-aminobutyric acid type A receptor subunit epsilon
- Gene Aliases
-
GABRG4
- Chromosome Location
-
Chr.4: 10827137 - 10858894 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205245.1 |
NP_990576.1 |
8 - 9 |
1164 |
99 |
GenBank mRNA |
X73533.1 |
- |
- |
1164 |
99 |
View Full Product Details