Assay Design



Gene Transcripts

Gene Symbol
CKB
Entrez Gene ID
396248
Gene Name
creatine kinase B
Gene Aliases
CPK-B
Chromosome Location
Chr.5: 50513988 - 50521688 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205310.1 NP_990641.1 5 - 6 816 128
XM_015287304.2 XP_015142790.1 6 - 7 843 128
XM_025150611.1 XP_025006379.1 7 - 8 1054 128
GenBank mRNA BX932948.1 - 6 - 7 827 128
X03509.1 - - 816 128

View Full Product Details