Assay Design



Gene Transcripts

Gene Symbol
ZPBP2
Entrez Gene ID
404532
Gene Name
zona pellucida binding protein 2
Gene Aliases
-
Chromosome Location
Chr.27: 7092628 - 7098535 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206987.1 NP_996870.1 3 - 4 344 115
XM_015299508.2 XP_015154994.1 3 - 4 1457 115
XM_025143967.1 XP_024999735.1 3 - 4 1457 115
GenBank mRNA AY575015.1 - 3 - 4 344 115

View Full Product Details