Assay Design



Gene Transcripts

Gene Symbol
UMOD
Entrez Gene ID
404754
Gene Name
uromodulin
Gene Aliases
ZPD
Chromosome Location
Chr.11: 523817 - 527808 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213576.2 NP_998741.2 2 - 3 95 65
XM_015292282.2 XP_015147768.1 1 - 2 66 65
GenBank mRNA AB114441.2 - 2 - 3 95 65

View Full Product Details