Assay Design



Gene Transcripts

Gene Symbol
NCK1
Entrez Gene ID
418727
Gene Name
NCK adaptor protein 1
Gene Aliases
NCK2
Chromosome Location
Chr.1: 136578811 - 136660607 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030814.1 NP_001025985.1 4 - 5 1219 100
GenBank mRNA AJ720236.1 - 4 - 5 1219 100

View Full Product Details