Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030917.1 | NP_001026088.1 | - | 2240 | 107 |
XM_025143694.1 | XP_024999462.1 | - | 1529 | 107 | |
XM_025143695.1 | XP_024999463.1 | - | 1521 | 107 | |
XM_025143696.1 | XP_024999464.1 | - | 647 | 107 | |
GenBank mRNA | AJ720521.1 | - | - | 2240 | 107 |