Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030934.1 | NP_001026105.1 | 3 - 4 | 443 | 72 |
XM_015299813.2 | XP_015155299.1 | 3 - 4 | 460 | 72 | |
GenBank mRNA | AJ719354.1 | - | 3 - 4 | 472 | 72 |
EF424417.2 | - | 3 - 4 | 416 | 72 |