Assay Design



Gene Transcripts

Gene Symbol
EHD3
Entrez Gene ID
421306
Gene Name
EH domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 8325103 - 8350291 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031040.1 NP_001026211.1 1 - 2 488 111
GenBank mRNA AJ719567.1 - 1 - 2 488 111

View Full Product Details