Assay Design



Gene Transcripts

Gene Symbol
RIC8A
Entrez Gene ID
422987
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
-
Chromosome Location
Chr.5: 1853549 - 1864662 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031174.1 NP_001026345.1 10 - 11 1659 84
XM_015286199.2 XP_015141685.1 10 - 11 1739 84
XM_025150649.1 XP_025006417.1 10 - 11 1712 84
GenBank mRNA AJ719857.1 - 10 - 11 1659 84

View Full Product Details