Assay Design



Gene Transcripts

Gene Symbol
PPIF
Entrez Gene ID
423723
Gene Name
peptidylprolyl isomerase F
Gene Aliases
-
Chromosome Location
Chr.6: 13095239 - 13103157 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031226.1 NP_001026397.1 3 - 4 334 149
GenBank mRNA AJ719394.1 - 3 - 4 334 149

View Full Product Details