Assay Design



Gene Transcripts

Gene Symbol
PMM2
Entrez Gene ID
427679
Gene Name
phosphomannomutase 2
Gene Aliases
-
Chromosome Location
Chr.14: 10454195 - 10461143 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031468.1 NP_001026639.1 4 - 5 376 107
GenBank mRNA AJ720584.1 - 4 - 5 376 107

View Full Product Details