Assay Design



Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
428752
Gene Name
amphiregulin
Gene Aliases
AREG/SDGF, AREGB
Chromosome Location
Chr.4: 44741666 - 44748282 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031537.1 NP_001026708.1 5 - 6 715 145
XM_015276315.2 XP_015131801.1 5 - 6 793 145
GenBank mRNA AY725836.1 - 5 - 6 715 145

View Full Product Details