Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030345.2 | NP_001025516.1 | 1 - 1 | 1050 | 66 |
XM_015279869.2 | XP_015135355.1 | 2 - 2 | 1258 | 66 | |
GenBank mRNA | AY954511.1 | - | 1 - 1 | 1050 | 66 |
DQ305354.1 | - | 1 - 1 | 1050 | 66 |