Assay Design



Gene Transcripts

Gene Symbol
TBX4
Entrez Gene ID
373989
Gene Name
T-box 4
Gene Aliases
-
Chromosome Location
Chr.19: 7734193 - 7760347 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030537.1 NP_001025708.1 3 - 4 357 62
GenBank mRNA AF033670.1 - 3 - 4 357 62
AF069395.1 - 1 - 2 54 62

View Full Product Details