Assay Design



Gene Transcripts

Gene Symbol
DPH1
Entrez Gene ID
417564
Gene Name
diphthamide biosynthesis 1
Gene Aliases
-
Chromosome Location
Chr.19: 5643569 - 5658310 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030716.2 NP_001025887.2 6 - 7 681 90
XM_015295755.2 XP_015151241.1 6 - 7 782 90
GenBank mRNA AJ721005.1 - 6 - 7 681 90

View Full Product Details