Assay Design



Gene Transcripts

Gene Symbol
DCN
Entrez Gene ID
417892
Gene Name
decorin
Gene Aliases
-
Chromosome Location
Chr.1: 44051182 - 44089820 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030747.2 NP_001025918.1 5 - 6 775 73
GenBank mRNA X63797.1 - - 778 73

View Full Product Details