Assay Design



Gene Transcripts

Gene Symbol
MLF2
Entrez Gene ID
418284
Gene Name
myeloid leukemia factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 77831588 - 77838988 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030776.2 NP_001025947.1 5 - 6 424 137
XM_015293367.2 XP_015148853.1 5 - 6 479 137
XM_015293370.2 XP_015148856.1 5 - 6 448 137
XM_015293372.2 XP_015148858.1 4 - 5 304 137
GenBank mRNA AJ851464.1 - 5 - 6 425 137

View Full Product Details