Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030894.2 | NP_001026065.1 | 2 - 3 | 322 | 62 |
XM_015297648.2 | XP_015153134.1 | 3 - 4 | 469 | 62 | |
GenBank mRNA | AJ719961.1 | - | 2 - 3 | 323 | 62 |
BX930386.1 | - | 2 - 3 | 305 | 62 | |
BX930954.1 | - | 3 - 4 | 399 | 62 | |
BX932588.1 | - | 2 - 3 | 306 | 62 | |
CR389031.1 | - | 2 - 3 | 303 | 62 |