Assay Design



Gene Transcripts

Gene Symbol
RPL11
Entrez Gene ID
419682
Gene Name
ribosomal protein L11
Gene Aliases
-
Chromosome Location
Chr.23: 5714104 - 5717202 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305188.1 NP_001292117.1 2 - 3 180 117
GenBank mRNA AJ720928.1 - 2 - 3 180 117

View Full Product Details