Assay Design



Gene Transcripts

Gene Symbol
METTL22
Entrez Gene ID
425865
Gene Name
methyltransferase like 22
Gene Aliases
C14H16orf68, C16orf68
Chromosome Location
Chr.14: 10509797 - 10520876 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278154.1 NP_001265083.1 5 - 6 899 134
GenBank mRNA AJ851587.1 - 5 - 6 897 134

View Full Product Details