Assay Design



Gene Transcripts

Gene Symbol
EOGT
Entrez Gene ID
426961
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Gene Aliases
AER61, AOS4, C12H3orf64, C3ORF64, EOGT1
Chromosome Location
Chr.12: 15674507 - 15690992 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031409.1 NP_001026580.1 2 - 3 318 59
XM_015293130.2 XP_015148616.1 3 - 4 563 59
XM_015293131.2 XP_015148617.1 3 - 4 435 59
XM_015293132.2 XP_015148618.1 3 - 4 565 59
GenBank mRNA AJ868233.1 - 2 - 3 318 59

View Full Product Details