Assay Design



Gene Transcripts

Gene Symbol
CNIH1
Entrez Gene ID
423561
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNI, CNIH
Chromosome Location
Chr.5: 56658853 - 56664637 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033006.1 NP_001028178.1 2 - 3 181 68
GenBank mRNA AB232678.1 - 2 - 3 181 68
BX929843.1 - 2 - 3 183 68
BX931023.1 - 2 - 3 232 68
BX933003.1 - 2 - 3 162 68
BX936003.1 - 2 - 3 167 68

View Full Product Details