Assay Design



Gene Transcripts

Gene Symbol
SFTPA1
Entrez Gene ID
423630
Gene Name
surfactant protein A1
Gene Aliases
LL
Chromosome Location
Chr.6: 5188498 - 5191583 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039166.1 NP_001034255.1 2 - 3 257 115
XM_025151510.1 XP_025007278.1 3 - 4 338 115
GenBank mRNA CR391234.1 - 2 - 3 256 115
DQ129667.1 - 2 - 3 257 115

View Full Product Details