Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277868.2 | NP_001264797.2 | 4 - 5 | 845 | 93 |
NM_001277869.2 | NP_001264798.1 | 4 - 5 | 572 | 93 | |
XM_025155467.1 | XP_025011235.1 | 4 - 5 | 558 | 93 | |
XM_025155468.1 | XP_025011236.1 | 4 - 5 | 609 | 93 | |
XM_025155469.1 | XP_025011237.1 | 4 - 5 | 473 | 93 | |
GenBank mRNA | AJ449366.1 | - | 4 - 5 | 541 | 93 |
AJ719514.1 | - | 4 - 5 | 536 | 93 | |
AJ719582.1 | - | 4 - 5 | 566 | 93 | |
AJ719663.1 | - | 4 - 5 | 534 | 93 | |
AJ720924.1 | - | - | 1171 | 93 | |
BX930410.1 | - | 4 - 5 | 845 | 93 | |
BX932518.2 | - | 4 - 5 | 845 | 93 | |
CN383670.1 | - | 4 - 5 | 536 | 93 |