Assay Design



Gene Transcripts

Gene Symbol
CORO1C
Entrez Gene ID
416893
Gene Name
coronin 1C
Gene Aliases
coronin-1C
Chromosome Location
Chr.15: 6830119 - 6868398 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277868.2 NP_001264797.2 8 - 9 1398 117
NM_001277869.2 NP_001264798.1 8 - 9 1125 117
XM_025155467.1 XP_025011235.1 8 - 9 1111 117
XM_025155468.1 XP_025011236.1 8 - 9 1162 117
XM_025155469.1 XP_025011237.1 8 - 9 1026 117
GenBank mRNA AJ720924.1 - - 1724 117
CO768688.1 - 4 - 5 517 117

View Full Product Details