Assay Design



Gene Transcripts

Gene Symbol
GUSB
Entrez Gene ID
427823
Gene Name
glucuronidase beta
Gene Aliases
beta-glucuronidase
Chromosome Location
Chr.19: 5065640 - 5081936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039316.2 NP_001034405.1 5 - 6 967 147
XM_015295810.2 XP_015151296.1 5 - 6 962 147
GenBank mRNA AJ720880.1 - 5 - 6 968 147

View Full Product Details