Assay Design
Gene Transcripts
- Gene Symbol
- RARRES1
- Entrez Gene ID
-
395209
- Gene Name
- retinoic acid receptor responder 1
- Gene Aliases
-
OCX32
- Chromosome Location
-
Chr.9: 22591970 - 22599460 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204534.4 |
NP_989865.1 |
3 - 4 |
487 |
116 |
GenBank mRNA |
AJ307060.2 |
- |
3 - 4 |
487 |
116 |
View Full Product Details