Assay Design



Gene Transcripts

Gene Symbol
TTR
Entrez Gene ID
396277
Gene Name
transthyretin
Gene Aliases
-
Chromosome Location
Chr.2: 106129430 - 106138303 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281498.1 NP_001268427.1 4 - 5 425 60
NM_205335.3 NP_990666.1 3 - 4 422 60
GenBank mRNA BU119521.1 - 3 - 4 364 60
BU269372.1 - 3 - 4 422 60
BU339400.1 - 3 - 4 364 60
BU352734.1 - 3 - 4 356 60
CR407350.1 - 4 - 5 425 60
DN851840.1 - 3 - 4 400 60
X60471.1 - - 364 60

View Full Product Details