Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039601.1 | NP_001034690.1 | 14 - 15 | 1508 | 87 |
XM_025152179.1 | XP_025007947.1 | 15 - 16 | 1562 | 87 | |
XM_025152180.1 | XP_025007948.1 | 12 - 13 | 1375 | 87 | |
GenBank mRNA | DQ150103.1 | - | 14 - 15 | 1508 | 87 |