Assay Design



Gene Transcripts

Gene Symbol
AOX2
Entrez Gene ID
424072
Gene Name
aldehyde oxidase 2
Gene Aliases
-
Chromosome Location
Chr.7: 11233181 - 11281017 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039601.1 NP_001034690.1 18 - 19 2047 75
XM_025152179.1 XP_025007947.1 19 - 20 2101 75
XM_025152180.1 XP_025007948.1 16 - 17 1914 75
GenBank mRNA DQ150103.1 - 18 - 19 2047 75

View Full Product Details