Assay Design



Gene Transcripts

Gene Symbol
AOX2
Entrez Gene ID
424072
Gene Name
aldehyde oxidase 2
Gene Aliases
-
Chromosome Location
Chr.7: 11233181 - 11281017 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039601.1 NP_001034690.1 25 - 26 2885 73
XM_025152179.1 XP_025007947.1 26 - 27 2939 73
XM_025152180.1 XP_025007948.1 23 - 24 2752 73
GenBank mRNA DQ150103.1 - 25 - 26 2885 73

View Full Product Details