Assay Design



Gene Transcripts

Gene Symbol
CTLA4
Entrez Gene ID
424106
Gene Name
cytotoxic T-lymphocyte associated protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 13409434 - 13412887 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040091.1 NP_001035180.1 2 - 3 542 64
GenBank mRNA AM236874.1 - 2 - 3 542 64

View Full Product Details