Assay Design



Gene Transcripts

Gene Symbol
SLC26A5
Entrez Gene ID
417715
Gene Name
solute carrier family 26 member 5
Gene Aliases
-
Chromosome Location
Chr.1: 13143551 - 13176906 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079477.1 NP_001072945.1 6 - 7 752 65
GenBank mRNA EF028087.1 - 6 - 7 752 65

View Full Product Details