Assay Design



Gene Transcripts

Gene Symbol
TMEM38A
Entrez Gene ID
771253
Gene Name
transmembrane protein 38A
Gene Aliases
-
Chromosome Location
Chr.28: 4384817 - 4391056 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079758.1 NP_001073226.1 4 - 5 594 145
XM_025144285.1 XP_025000053.1 5 - 6 661 145
GenBank mRNA AJ720241.1 - 4 - 5 594 145

View Full Product Details