Assay Design



Gene Transcripts

Gene Symbol
TAS2R7
Entrez Gene ID
771663
Gene Name
taste receptor, type 2, member 7
Gene Aliases
Tas2r3
Chromosome Location
Chr.3: 109851221 - 109852186 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080719.1 NP_001074188.1 1 - 1 840 98

View Full Product Details