Assay Design



Gene Transcripts

Gene Symbol
SHFM1
Entrez Gene ID
772237
Gene Name
split hand/foot malformation (ectrodactyly) type 1
Gene Aliases
C7orf76, SEM1
Chromosome Location
Chr.2: 24321442 - 24327977 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085439.1 NP_001078908.1 1 - 2 158 68
GenBank mRNA AJ720317.1 - 1 - 2 156 68
BX950613.1 - 1 - 2 127 68

View Full Product Details