Assay Design



Gene Transcripts

Gene Symbol
MT3
Entrez Gene ID
770592
Gene Name
metallothionein 3
Gene Aliases
MT1
Chromosome Location
Chr.11: 2122326 - 2123174 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097538.1 NP_001091007.1 1 - 2 33 131

View Full Product Details