Assay Design
Gene Transcripts
- Gene Symbol
- GUCA1A
- Entrez Gene ID
-
374216
- Gene Name
- guanylate cyclase activator 1A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.26: 3184817 - 3189219 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204320.2 |
NP_989651.1 |
- |
468 |
73 |
GenBank mRNA |
S82199.1 |
- |
- |
468 |
73 |
View Full Product Details